Assay Details
Target Gene Details
Entrez Gene ID: | 18429 |
Gene Name: | oxytocin |
Gene Aliases: |
OT, Oxy |
Location: |
Chr.2:130574967-130577054 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Oxt | NM_011025.4 | 3 | 414 | NP_035155.1 |
| AF483508.1 | AAL90782.1 | |||
| AF483509.1 | AAL90783.1 | |||
| AK003042.1 | 3 | 411 | BAB22527.1 | |
| AW455577.1 | 1 | 142 | ||
| BC117030.1 | 3 | 393 | ||
| BC117032.1 | 3 | 393 |
More Information
Set Membership: |
|

Genomic Map