Assay Details
Target Gene Details
Entrez Gene ID: | 100502946 |
Gene Name: | predicted gene, 16796 |
Gene Aliases: |
- |
Location: |
Chr.2:170499357-170511927 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm16796 | NR_040367.1 | 1 | 584 | |
| AK138533.1 | 1 | 585 |
Target Gene Details
Entrez Gene ID: | 109054 |
Gene Name: | prefoldin 4 |
Gene Aliases: |
0610025M10Rik, 1810063B06Rik, C-1, C1, Pfd4 |
Location: |
Chr.2:170496428-170519070 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfdn4 | NM_001013369.2 | NP_001013387.1 | ||
| NM_001110152.2 | 1 | 115 | NP_001103622.1 | |
| NM_001199902.1 | 1 | 115 | NP_001186831.1 | |
| AK136249.1 | 1 | 116 | BAE22896.1 | |
| BC065407.1 | AAH65407.1 | |||
| BC147673.1 | ||||
| BC147674.1 |

Genomic Map