Assay Details
Target Gene Details
Entrez Gene ID: | 102238433 |
Gene Name: | predicted gene, 28042 |
Gene Aliases: |
Jmjd7-pla2g4b |
Location: |
Chr.2:120027483-120043033 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm28042 | NR_104353.1 | |||
| AB195274.1 | ||||
| AB195275.1 |
Target Gene Details
Entrez Gene ID: | 211429 |
Gene Name: | phospholipase A2, group IVB (cytosolic) |
Gene Aliases: |
A030011C02Rik, Gm1013, cPLA2-beta |
Location: |
Chr.2:120033433-120043033 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g4b | NM_145378.4 | NP_663353.3 | ||
| AK037218.1 | ||||
| AK137721.1 | ||||
| BC042758.1 | AAH42758.1 | |||
| DQ888308.1 |
More Information
Set Membership: |
|

Genomic Map