Assay Details
Target Gene Details
Entrez Gene ID: | 16922 |
Gene Name: | phytanoyl-CoA hydroxylase |
Gene Aliases: |
AI256161, AI265699, Lnap1, PAHX |
Location: |
Chr.2:4918996-4938743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phyh | NM_010726.2 | NP_034856.1 | ||
| AF023463.1 | AAB81835.1 | |||
| AK146753.1 | BAE27410.1 | |||
| AK151372.1 | ||||
| AK164495.1 | BAE37810.1 | |||
| AK165412.1 | BAE38170.1 | |||
| AK168648.1 | BAE40505.1 | |||
| AK169213.1 | BAE40985.1 | |||
| BC002018.1 | AAH02018.1 | |||
| D88670.1 | BAA19003.1 |
More Information
Set Membership: |
|

Genomic Map