Assay Details
Target Gene Details
Entrez Gene ID: | 228812 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class U |
Gene Aliases: |
5430426F17Rik, BC028278, Cdc91l1 |
Location: |
Chr.2:155278243-155357424 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigu | NM_001004721.1 | NP_001004721.1 | ||
| AK143517.1 | BAE25411.1 | |||
| AK171988.1 | BAE42761.1 | |||
| AK211799.1 | ||||
| AW046539.1 | ||||
| BC120680.1 | ||||
| BC120682.1 | ||||
| CK334159.1 |

Genomic Map