Assay Details
Target Gene Details
Entrez Gene ID: | 228852 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 16B |
Gene Aliases: |
ANKRD4, C130078N17Rik, TIMAP, Wdt4 |
Location: |
Chr.2:158666733-158766334 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r16b | NM_001159662.1 | NP_001153134.1 | ||
| NM_153089.4 | NP_694729.1 | |||
| AF423761.1 | AAL62093.1 | |||
| AK042123.1 | BAC31176.1 | |||
| AK044453.1 | ||||
| BC054764.1 | AAH54764.1 | |||
| BC057542.1 | AAH57542.1 |
More Information
Set Membership: |
|

Genomic Map