Assay Details
Target Gene Details
Entrez Gene ID: | 18803 |
Gene Name: | phospholipase C, gamma 1 |
Gene Aliases: |
AI894140, Cded, Plc-1, Plc-gamma1, Plcg-1 |
Location: |
Chr.2:160731310-160775760 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcg1 | NM_021280.3 | 20 | 2297 | NP_067255.2 |
| AK134041.1 | 19 | 2102 | BAE21990.1 | |
| AK162809.1 | 8 | 969 | ||
| BC065091.1 | 20 | 2297 | AAH65091.1 |
More Information
Set Membership: |
|

Genomic Map