Assay Details
Target Gene Details
Entrez Gene ID: | 241274 |
Gene Name: | patatin-like phospholipase domain containing 7 |
Gene Aliases: |
BC027342, E430013P11Rik, Nre |
Location: |
Chr.2:24975171-25054072 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpla7 | NM_146251.4 | 23 | 2698 | NP_666363.3 |
| AK088362.1 | 23 | 2644 | BAC40302.1 | |
| AK148380.1 | 23 | 2672 | BAE28519.1 | |
| AK170417.1 | 23 | 2644 | BAE41783.1 | |
| BC025621.1 | 19 | 2111 | AAH25621.1 | |
| BC027342.1 | 1 | 142 | AAH27342.1 | |
| BC064003.1 | 6 | 462 | AAH64003.1 | |
| BC080793.1 | AAH80793.1 | |||
| BC141365.1 | 23 | 2482 | ||
| BC141366.1 | 23 | 2482 | ||
| EU046569.1 | 3 | 259 | ||
| EU293208.1 | 22 | 2369 | ||
| EU368675.1 | 22 | 2369 | ||
| EU402948.2 | 21 | 2305 |
More Information
Set Membership: |
|

Genomic Map