Assay Details
Target Gene Details
Entrez Gene ID: | 227700 |
Gene Name: | SH3-domain GRB2-like endophilin B2 |
Gene Aliases: |
SPAS-1 |
Location: |
Chr.2:30344777-30359316 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3glb2 | NM_001289709.1 | NP_001276638.1 | ||
| NM_001289710.1 | NP_001276639.1 | |||
| NM_139302.2 | NP_647463.1 | |||
| AF426314.1 | AAL24820.1 | |||
| AK129455.1 | BAC98265.2 | |||
| AK152409.1 | BAE31195.1 | |||
| AK152502.1 | ||||
| AK155248.1 | BAE33146.1 | |||
| AK167762.1 | BAE39795.1 | |||
| AK182931.1 | ||||
| AK188040.1 | ||||
| BC024477.1 | AAH24477.1 | |||
| BC028859.1 | AAH28859.1 | |||
| BY317126.1 | ||||
| CX216029.1 | ||||
| EF676083.1 |

Genomic Map