Assay Details
Target Gene Details
Entrez Gene ID: | 20515 |
Gene Name: | solute carrier family 20, member 1 |
Gene Aliases: |
AI607883, Glvr-1, Glvr1 |
Location: |
Chr.2:129198773-129211615 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc20a1 | NM_001159593.1 | 1 | 69 | NP_001153065.1 |
| NM_015747.2 | 1 | 69 | NP_056562.1 | |
| AK003059.1 | BAC25018.1 | |||
| AK035898.1 | BAC29234.1 | |||
| AK084159.1 | ||||
| AK089184.1 | ||||
| AK155506.1 | BAE33298.1 | |||
| CD545381.1 | 1 | 69 |
More Information
Set Membership: |
|

Genomic Map