Assay Details
Target Gene Details
Entrez Gene ID: | 24105 |
Gene Name: | RanBP-type and C3HC4-type zinc finger containing 1 |
Gene Aliases: |
AL033326, HOIL-1, HOIL-1L, UIP28, Ubce7ip3 |
Location: |
Chr.2:152316334-152332643 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbck1 | NM_019705.3 | 1 | 71 | NP_062679.2 |
| BC034555.1 | 1 | 67 | AAH34555.1 | |
| BY084037.1 | 1 | 85 | ||
| BY267597.1 | 1 | 50 |
More Information
Set Membership: |
|

Genomic Map