Assay Details
Target Gene Details
Entrez Gene ID: | 24105 |
Gene Name: | RanBP-type and C3HC4-type zinc finger containing 1 |
Gene Aliases: |
AL033326, HOIL-1, HOIL-1L, UIP28, Ubce7ip3 |
Location: |
Chr.2:152316334-152332643 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbck1 | NM_001083921.1 | NP_001077390.1 | ||
| NM_019705.3 | NP_062679.2 | |||
| AF124663.1 | AAD24572.1 | |||
| AK085559.1 | ||||
| AK088591.1 | BAC40440.1 | |||
| AK146176.1 | ||||
| AK166075.1 | BAE38556.1 | |||
| AK209721.1 | ||||
| BC034555.1 | AAH34555.1 |
More Information
Set Membership: |
|

Genomic Map