Assay Details
Target Gene Details
Entrez Gene ID: | 20638 |
Gene Name: | small nuclear ribonucleoprotein B |
Gene Aliases: |
AL024368, AU018828, SM-B, SM11, SMB, SNRNP-B |
Location: |
Chr.2:130171636-130179376 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snrpb | NM_009225.2 | 1 | 69 | NP_033251.1 |
| AK077537.1 | ||||
| AK146319.1 | 1 | 112 | BAE27074.1 | |
| BG086527.2 | ||||
| BY280308.1 | 1 | 69 |
More Information
Set Membership: |
|

Genomic Map