Assay Details
Target Gene Details
Entrez Gene ID: | 20272 |
Gene Name: | sodium channel, voltage-gated, type VII, alpha |
Gene Aliases: |
1110034K09Rik, AI642000, NaG, Nav2, Nav2.3, Nax, Scn6a |
Location: |
Chr.2:66673424-66785091 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn7a | NM_009135.2 | NP_033161.2 | ||
| L36179.1 | AAA66192.1 |
More Information
Set Membership: |
|

Genomic Map