Assay Details
Target Gene Details
Entrez Gene ID: | 94229 |
Gene Name: | solute carrier family 4, sodium bicarbonate cotransporter-like, member 10 |
Gene Aliases: |
NCBE, mKIAA4136 |
Location: |
Chr.2:62046515-62326743 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a10 | NM_001242378.1 | 22 | 3081 | NP_001229307.1 |
| NM_001242379.1 | 21 | 2991 | NP_001229308.1 | |
| NM_001242380.1 | 22 | 3081 | NP_001229309.1 | |
| NM_001242381.1 | 21 | 2988 | NP_001229310.1 | |
| NM_001242382.1 | 22 | 3078 | NP_001229311.1 | |
| NM_001242383.1 | 21 | 2988 | NP_001229312.1 | |
| NM_033552.3 | 21 | 2991 | NP_291030.2 | |
| AB033759.1 | 21 | 2912 | BAB17922.1 | |
| AK043017.1 | 21 | 2967 | BAC31434.2 | |
| AK220501.1 | 22 | 3046 | BAD90511.1 | |
| BC039226.1 | 21 | 2940 | AAH39226.1 | |
| HM149278.1 | 21 | 2952 | ||
| HQ533147.1 | 22 | 3018 | ||
| HQ825316.1 | 22 | 3021 | ||
| HQ831517.1 | 21 | 2928 | ||
| JF500487.1 | 21 | 3068 | ||
| JF500488.1 | 22 | 3158 | ||
| JF500489.1 | 21 | 3068 | ||
| JF500490.1 | 22 | 3158 | ||
| JX220977.1 | 22 | 3045 | ||
| JX254917.1 | 21 | 2958 | ||
| JX268544.1 | 22 | 3159 |
More Information
Set Membership: |
|

Genomic Map