Assay Details
Target Gene Details
Entrez Gene ID: | 269356 |
Gene Name: | solute carrier family 4, sodium bicarbonate transporter-like, member 11 |
Gene Aliases: |
AI503023, BTR1, NaBC1 |
Location: |
Chr.2:130684107-130697519 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a11 | NM_001081162.1 | NP_001074631.1 | ||
| AK141286.1 | BAE24637.1 | |||
| AK158115.1 | ||||
| BC111884.1 |
More Information
Set Membership: |
|

Genomic Map