Assay Details
Target Gene Details
Entrez Gene ID: | 110078 |
Gene Name: | brain glycogen phosphorylase |
Gene Aliases: |
- |
Location: |
Chr.2:150786796-150831748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pygb | NM_153781.1 | NP_722476.1 | ||
| AK032769.1 | BAE20482.1 | |||
| AK035673.1 | ||||
| AK133794.1 | BAE21845.1 | |||
| AK134667.1 | BAE22233.1 | |||
| AK142716.1 | ||||
| AK147763.1 | BAE28122.1 | |||
| AK168381.1 | ||||
| AK169052.1 | BAE40840.1 | |||
| AK169631.1 | ||||
| AK177572.1 | ||||
| AK185990.1 | ||||
| BC032209.1 | AAH32209.1 | |||
| BC035283.1 | AAH35283.1 |
More Information
Set Membership: |
|

Genomic Map