Assay Details
Target Gene Details
Entrez Gene ID: | 20274 |
Gene Name: | sodium channel, voltage-gated, type IX, alpha |
Gene Aliases: |
Nav1.7, PN1, mKIAA4197 |
Location: |
Chr.2:66480080-66635345 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn9a | NM_001290674.1 | NP_001277603.1 | ||
| NM_001290675.1 | NP_001277604.1 | |||
| AK141871.1 | ||||
| AK142068.1 | BAE24929.1 | |||
| BC158048.1 | ||||
| BC172147.1 |
More Information
Set Membership: |
|

Genomic Map