Assay Details
Target Gene Details
Entrez Gene ID: | 269356 |
Gene Name: | solute carrier family 4, sodium bicarbonate transporter-like, member 11 |
Gene Aliases: |
AI503023, BTR1, NaBC1 |
Location: |
Chr.2:130684107-130697519 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc4a11 | NM_001081162.1 | 9 | 1049 | NP_001074631.1 |
| AK052665.1 | 5 | 1845 | ||
| AK158115.1 | 9 | 1003 | ||
| BC111884.1 | 8 | 844 |
More Information
Set Membership: |
|

Genomic Map