Assay Details
Target Gene Details
Entrez Gene ID: | 99138 |
Gene Name: | START domain containing 7 |
Gene Aliases: |
AI852671, AL022671, AW544915 |
Location: |
Chr.2:127270203-127298935 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stard7 | NM_139308.2 | 2 | 793 | NP_647469.2 |
| AF244543.1 | 1 | 150 | ||
| AK044519.1 | 2 | 289 | BAC31961.1 | |
| AK083125.1 | 2 | 320 | BAC38772.1 | |
| AK143699.1 | 2 | 794 | BAE25505.1 | |
| BC017524.1 | 2 | 783 | ||
| BC080747.1 | 2 | 764 | AAH80747.1 |

Genomic Map