Assay Details
Target Gene Details
Entrez Gene ID: | 72043 |
Gene Name: | sulfatase 2 |
Gene Aliases: |
2010004N24Rik, AU020235, MSulf-2, mKIAA1247 |
Location: |
Chr.2:166073899-166155683 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sulf2 | NM_001252578.1 | NP_001239507.1 | ||
| NM_001252579.1 | NP_001239508.1 | |||
| NM_028072.5 | NP_082348.2 | |||
| AK008108.1 | BAB25464.1 | |||
| AK028874.1 | BAC26165.1 | |||
| AK034712.1 | BAC28804.2 | |||
| AK036685.1 | BAC29534.1 | |||
| AK049170.1 | BAC33584.1 | |||
| AK081643.1 | BAC38279.1 | |||
| AK129316.1 | BAC98126.1 | |||
| AK165183.1 | BAE38065.1 | |||
| AY101177.1 | AAM76862.1 | |||
| BC141086.1 |
More Information
Set Membership: |
|

Genomic Map