Assay Details
Target Gene Details
Entrez Gene ID: | 277360 |
Gene Name: | phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1 |
Gene Aliases: |
G630042G04, P-REX1, Setd6 |
Location: |
Chr.2:166566345-166713832 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prex1 | NM_177782.3 | 17 | 2030 | NP_808450.2 |
| AK173168.1 | 17 | 2030 | BAD32446.1 | |
| BC151074.1 | 17 | 1978 | ||
| BC157940.1 | 17 | 1978 |
More Information
Set Membership: |
|

Genomic Map