Assay Details
Target Gene Details
Entrez Gene ID: | 76938 |
Gene Name: | RNA binding motif protein 17 |
Gene Aliases: |
2700027J02Rik |
Location: |
Chr.2:11585439-11603199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm17 | NM_152824.1 | NP_690037.1 | ||
| AF083384.1 | AAC64085.1 | |||
| AK012296.1 | ||||
| AK180353.1 | ||||
| BC034896.1 | AAH34896.1 |
More Information
Set Membership: |
|

Genomic Map