Assay Details
Target Gene Details
Entrez Gene ID: | 76857 |
Gene Name: | speckle-type POZ protein-like |
Gene Aliases: |
4921517N04Rik, AU014935, BB233739, E430033K04Rik |
Location: |
Chr.2:23506220-23572128 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spopl | NM_001165997.1 | NP_001159469.1 | ||
| NM_001165998.1 | NP_001159470.1 | |||
| NM_029773.2 | NP_084049.2 | |||
| AK162056.1 | BAE36701.1 | |||
| BC111867.1 | AAI11868.1 | |||
| BI648825.1 | ||||
| CF746115.1 |
More Information
Set Membership: |
|

Genomic Map