Assay Details
Target Gene Details
Entrez Gene ID: | 277360 |
Gene Name: | phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 1 |
Gene Aliases: |
G630042G04, P-REX1, Setd6 |
Location: |
Chr.2:166566345-166713832 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prex1 | NM_177782.3 | NP_808450.2 | ||
| AK090301.1 | BAC41161.1 | |||
| AK173168.1 | BAD32446.1 | |||
| BC057617.1 | AAH57617.1 | |||
| BC151074.1 | ||||
| BC157940.1 |
More Information
Set Membership: |
|

Genomic Map