Assay Details
Target Gene Details
Entrez Gene ID: | 56086 |
Gene Name: | SET nuclear oncogene |
Gene Aliases: |
2610030F17Rik, 5730420M11Rik, AA407739, I-2PP2A, StF-IT-1, TAF-I |
Location: |
Chr.2:30061996-30072577 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Set | NM_023871.4 | 1 | 87 | NP_076360.1 |
| AB044937.1 | BAB20793.1 | |||
| AK014291.1 | 1 | 205 | BAB29248.3 | |
| AK031561.1 | 1 | 203 | ||
| AK054400.1 | 1 | 310 | ||
| AK172327.1 | 1 | 231 | BAE42947.1 | |
| BC018255.1 | 1 | 40 | AAH18255.1 | |
| BE331951.1 | 1 | 87 |
More Information
Set Membership: |
|

Genomic Map