Assay Details
Target Gene Details
Entrez Gene ID: | 20732 |
Gene Name: | serine protease inhibitor, Kunitz type 1 |
Gene Aliases: |
HAI-1 |
Location: |
Chr.2:119237360-119249535 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spint1 | NM_016907.3 | NP_058603.2 | ||
| AF099018.1 | AAF02490.1 | |||
| AK017342.1 | BAB30697.1 | |||
| BC005769.1 | AAH05769.1 |
More Information
Set Membership: |
|

Genomic Map