Assay Details
Target Gene Details
Entrez Gene ID: | 53416 |
Gene Name: | serine/threonine kinase 39 |
Gene Aliases: |
AW227544, AW556857, DCHT, RF005, Rnl5, SPAK |
Location: |
Chr.2:68210445-68472965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stk39 | NM_016866.2 | NP_058562.1 | ||
AF099988.1 | AAC72237.1 | |||
AK017292.1 | ||||
AK148456.1 | ||||
BC051964.1 | AAH51964.2 | |||
BC064443.1 | AAH64443.1 | |||
JN368425.1 |
