Assay Details
Target Gene Details
Entrez Gene ID: | 320558 |
Gene Name: | synaptonemal complex protein 2 |
Gene Aliases: |
3830402K23Rik, 4930518F03Rik |
Location: |
Chr.2:178345295-178407658 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sycp2 | NM_177191.3 | NP_796165.2 | ||
| AK036624.1 | ||||
| DQ103262.1 |
More Information
Set Membership: |
|

Genomic Map