Assay Details
Target Gene Details
Entrez Gene ID: | 23992 |
Gene Name: | protein kinase, interferon inducible double stranded RNA dependent activator |
Gene Aliases: |
AV120107, PRK, Pact, RAX, lear |
Location: |
Chr.2:76609215-76648199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkra | NM_011871.2 | NP_036001.1 | ||
| AF083032.1 | AAD33098.1 | |||
| AK012798.1 | BAB28477.1 | |||
| AK037072.1 | 1 | 1102 | ||
| AK155112.1 | BAE33056.1 | |||
| BC011311.1 | AAH11311.1 |

Genomic Map