Assay Details
Target Gene Details
Entrez Gene ID: | 19215 |
Gene Name: | prostaglandin D2 synthase (brain) |
Gene Aliases: |
21kDa, L-PGDS, PGD2, PGDS, PGDS2, Ptgs3 |
Location: |
Chr.2:25466709-25470109 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgds | NM_008963.2 | NP_032989.2 | ||
| AB006361.1 | BAA21769.1 | |||
| AK002382.1 | ||||
| AK131859.1 | BAE20833.1 | |||
| BC038083.1 | 5 | 826 | AAH38083.1 | |
| BC043015.1 |
More Information
Set Membership: |
|

Genomic Map