Assay Details
Target Gene Details
Entrez Gene ID: | 227800 |
Gene Name: | RAB GTPase activating protein 1 |
Gene Aliases: |
Gapcena, mKIAA4104 |
Location: |
Chr.2:37443261-37566454 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabgap1 | NM_001033960.1 | 11 | 1549 | NP_001029132.1 |
| NM_146121.2 | 11 | 1673 | NP_666233.2 | |
| AK029667.1 | 11 | 1535 | ||
| AK044346.1 | 11 | 1550 | BAC31880.1 | |
| AK220489.1 | 10 | 1494 | BAD90509.1 | |
| BC138337.1 | 11 | 1575 |
More Information
Set Membership: |
|

Genomic Map