Assay Details
Target Gene Details
Entrez Gene ID: | 228875 |
Gene Name: | solute carrier family 35, member C2 |
Gene Aliases: |
C85957, CGI-15, D2Wsu58e, Ovcov1 |
Location: |
Chr.2:165276522-165287888 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35c2 |

Genomic Map