Assay Details
Target Gene Details
Entrez Gene ID: | 20740 |
Gene Name: | spectrin alpha, non-erythrocytic 1 |
Gene Aliases: |
2610027H02Rik, Spna-2, Spna2 |
Location: |
Chr.2:29965553-30031451 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptan1 | NM_001177667.1 | NP_001171138.1 | ||
| NM_001177668.1 | NP_001171139.1 | |||
| AK147317.1 | ||||
| AK164891.1 | BAE37955.1 | |||
| BC027791.1 | AAH27791.1 | |||
| BC150941.1 | ||||
| BC158015.1 | ||||
| BC172095.1 | ||||
| X12801.1 | CAA31289.1 |

Genomic Map