Assay Details
Target Gene Details
Entrez Gene ID: | 320558 |
Gene Name: | synaptonemal complex protein 2 |
Gene Aliases: |
3830402K23Rik, 4930518F03Rik |
Location: |
Chr.2:178345295-178407658 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sycp2 | NM_177191.3 | 30 | 2907 | NP_796165.2 |
| AJ409504.1 | 2 | 143 | ||
| AK029735.1 | 3 | 218 | BAC26589.1 | |
| AK036624.1 | 8 | 755 | ||
| DQ103262.1 | 30 | 2907 |
More Information
Set Membership: |
|

Genomic Map