Assay Details
Target Gene Details
Entrez Gene ID: | 56508 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 4 |
Gene Aliases: |
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040 |
Location: |
Chr.2:71981215-72257474 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef4 | NM_001204165.1 | NP_001191094.1 | ||
| NM_001204166.1 | NP_001191095.1 | |||
| NM_019688.2 | NP_062662.1 | |||
| AB021132.1 | BAB18976.1 | |||
| AB037668.1 | BAB72180.1 | |||
| AB445011.1 | ||||
| AF115480.1 | AAD09132.1 | |||
| AK004874.1 | BAB23633.1 | |||
| AK203721.1 | ||||
| AK220281.1 | BAD90206.1 | |||
| BC139822.1 |
More Information
Set Membership: |
|

Genomic Map