Assay Details
Target Gene Details
Entrez Gene ID: | 114644 |
Gene Name: | solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3 |
Gene Aliases: |
NaDC-3, NaDC3, SDCT2 |
Location: |
Chr.2:165405295-165473225 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a3 | NM_054055.2 | NP_473396.1 | ||
| AF306491.1 | AAK49898.1 | |||
| AK085326.1 | ||||
| AK086775.1 | ||||
| AK087518.1 | ||||
| AK128982.1 | ||||
| AK138344.1 | BAE23629.1 | |||
| AK157881.1 | ||||
| AK165355.1 | ||||
| BC026803.1 | AAH26803.1 |
More Information
Set Membership: |
|

Genomic Map