Assay Details
Target Gene Details
Entrez Gene ID: | 21969 |
Gene Name: | topoisomerase (DNA) I |
Gene Aliases: |
AI467334, D130064I21Rik, Top-1 |
Location: |
Chr.2:160645897-160722764 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Top1 | NM_009408.2 | 12 | 1403 | NP_033434.2 |
| CO425650.1 | 2 | 208 | ||
| D10061.1 | 12 | 1070 | BAA00950.1 | |
| L20632.1 | 12 | 1067 | AAA40466.1 |
More Information
Set Membership: |
|

Genomic Map