Assay Details
Target Gene Details
Entrez Gene ID: | 72147 |
Gene Name: | zinc finger and BTB domain containing 46 |
Gene Aliases: |
2610019F01Rik, 4933406L05Rik, BZEL, Btbd4 |
Location: |
Chr.2:181371261-181492982 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb46 |
Target Gene Details
Entrez Gene ID: | 229007 |
Gene Name: | zinc finger, CCCH-type with G patch domain |
Gene Aliases: |
1500006I01Rik, BC021513 |
Location: |
Chr.2:181364931-181380793 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zgpat | NM_001048148.1 | 4 | 1182 | NP_001041613.1 |
| NM_144894.3 | 5 | 1406 | NP_659143.1 | |
| AK041491.1 | 5 | 1410 | BAC30962.1 | |
| AK049764.1 | 4 | 1181 | BAC33909.1 | |
| AK052236.1 | 4 | 1080 | BAC34894.1 | |
| AK159801.1 | 4 | 1201 | BAE35381.1 | |
| AK209697.1 | 3 | 259 | ||
| BC021513.1 | 4 | 1180 | AAH21513.1 | |
| BC027218.1 | 4 | 1089 | AAH27218.1 |

Genomic Map