Assay Details
Target Gene Details
Entrez Gene ID: | 320799 |
Gene Name: | zinc fingers and homeoboxes 3 |
Gene Aliases: |
1810059C13Rik, 4932418O04Rik, 9530010N21Rik, Tix1, mKIAA0395 |
Location: |
Chr.2:160770447-160872990 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zhx3 | NM_177263.3 | 1 | 59 | NP_796237.2 |
| AK143296.1 | 1 | 40 | BAE25337.1 | |
| BC058111.1 | 1 | 59 | AAH58111.1 |
More Information
Set Membership: |
|

Genomic Map