Assay Details
Target Gene Details
Entrez Gene ID: | 21353 |
Gene Name: | TRAF family member-associated Nf-kappa B activator |
Gene Aliases: |
C86182, E430026L09Rik, I-TRAF |
Location: |
Chr.2:61578561-61654171 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tank | NM_001164071.1 | 3 | 354 | NP_001157543.1 |
| NM_001164072.1 | 2 | 149 | NP_001157544.1 | |
| NM_011529.2 | 2 | 145 | NP_035659.1 | |
| AK031559.1 | 2 | 111 | ||
| AK038910.1 | 2 | 84 | ||
| AK088817.1 | 2 | 95 | BAC40592.1 | |
| AK147727.1 | 3 | 300 | BAE28097.1 | |
| AK156658.1 | ||||
| AK165420.1 | 2 | 105 | BAE38175.1 | |
| AK165994.1 | 2 | 80 | ||
| BB660164.1 | 2 | 150 | ||
| BC026148.1 | 2 | 130 | AAH26148.1 | |
| BC033282.1 | 2 | 81 | ||
| U51907.1 | 2 | 87 | AAB02204.1 | |
| U59864.1 | 2 | 158 | AAC52781.1 |

Genomic Map