Assay Details
Target Gene Details
Entrez Gene ID: | 21353 |
Gene Name: | TRAF family member-associated Nf-kappa B activator |
Gene Aliases: |
C86182, E430026L09Rik, I-TRAF |
Location: |
Chr.2:61578561-61654171 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tank | NM_001164071.1 | NP_001157543.1 | ||
| NM_001164072.1 | NP_001157544.1 | |||
| NM_011529.2 | NP_035659.1 | |||
| AK031559.1 | ||||
| AK088817.1 | BAC40592.1 | |||
| AK147727.1 | BAE28097.1 | |||
| AK165420.1 | BAE38175.1 | |||
| AK165994.1 | ||||
| BB660164.1 | ||||
| BC026148.1 | AAH26148.1 | |||
| BC033282.1 | ||||
| U51907.1 | AAB02204.1 | |||
| U59864.1 | AAC52781.1 |

Genomic Map