Assay Details
Target Gene Details
Entrez Gene ID: | 228880 |
Gene Name: | zinc finger, MYND-type containing 8 |
Gene Aliases: |
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7 |
Location: |
Chr.2:165784152-165898984 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmynd8 | NM_001252584.2 | 19 | 3162 | NP_001239513.1 |
| NM_001252585.1 | 18 | 3054 | NP_001239514.1 | |
| NM_001281926.1 | 19 | 3061 | NP_001268855.1 | |
| NM_001291158.1 | 20 | 5327 | NP_001278087.1 | |
| NM_027230.5 | 20 | 3405 | NP_081506.3 | |
| AB093284.2 | 20 | 5319 | BAC41468.4 | |
| AK014397.1 | 7 | 1462 | BAE43241.1 | |
| AK132427.1 | 18 | 3056 | BAE21162.1 | |
| AK147620.1 | 19 | 3062 | BAE28029.1 | |
| AK155864.1 | 19 | 2967 | BAE33468.1 | |
| BC041797.1 | 9 | 2133 | AAH41797.1 | |
| BC048186.1 | 20 | 3212 | AAH48186.3 |

Genomic Map