Assay Details
Target Gene Details
Entrez Gene ID: | 192140 |
Gene Name: | transmembrane channel-like gene family 2 |
Gene Aliases: |
CWEA2 |
Location: |
Chr.2:130195194-130264445 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmc2 | NM_138655.1 | 2 | 236 | NP_619596.1 |
| AF417581.1 | 2 | 236 | AAL86402.1 |

Genomic Map