Assay Details
Target Gene Details
Entrez Gene ID: | 11532 |
Gene Name: | alcohol dehydrogenase 5 (class III), chi polypeptide |
Gene Aliases: |
Adh-5, Adh3, GSNOR |
Location: |
Chr.3:138437200-138455499 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh5 | NM_001288578.1 | 5 | 565 | NP_001275507.1 |
| NM_007410.3 | 5 | 583 | NP_031436.2 | |
| AK002790.1 | 5 | 505 | ||
| AK076507.1 | 5 | 512 | BAC36370.1 | |
| AK146949.1 | 5 | 527 | BAE27558.1 | |
| AK159803.1 | 5 | 563 | BAE35383.1 | |
| AK214149.1 | 1 | 42 | ||
| BC062879.1 | 5 | 509 | ||
| BC090978.1 | 5 | 506 | AAH90978.1 | |
| CF172597.1 | 5 | 565 | ||
| M84147.1 | 5 | 533 | AAA68896.1 |
More Information
Set Membership: |
|

Genomic Map