Assay Details
Target Gene Details
Entrez Gene ID: | 11532 |
Gene Name: | alcohol dehydrogenase 5 (class III), chi polypeptide |
Gene Aliases: |
Adh-5, Adh3, GSNOR |
Location: |
Chr.3:138437200-138455499 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh5 | NM_001288578.1 | NP_001275507.1 | ||
| NM_007410.3 | NP_031436.2 | |||
| AK002790.1 | ||||
| AK076507.1 | BAC36370.1 | |||
| AK146949.1 | BAE27558.1 | |||
| AK159803.1 | BAE35383.1 | |||
| BC062879.1 | ||||
| BC090978.1 | AAH90978.1 | |||
| M84147.1 | AAA68896.1 |
More Information
Set Membership: |
|

Genomic Map