Assay Details
Target Gene Details
Entrez Gene ID: | 69117 |
Gene Name: | alcohol dehydrogenase 6A (class V) |
Gene Aliases: |
1810009C16Rik, Adh5a |
Location: |
Chr.3:138313286-138331134 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adh6a | NM_026945.1 | 4 | 364 | NP_081221.1 |
AK007397.1 | 4 | 360 | BAB25012.1 | |
AK028114.1 | 4 | 365 | ||
BC130016.1 | 4 | 333 |
