Assay Details
Target Gene Details
Entrez Gene ID: | 74270 |
Gene Name: | ubiquitin specific peptidase 20 |
Gene Aliases: |
1700055M05Rik, AI467231, Vdu2 |
Location: |
Chr.2:30982279-31022655 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp20 | NM_028846.5 | NP_083122.1 | ||
| AF449715.1 | AAN15803.1 | |||
| AK042612.1 | BAC31303.1 | |||
| AK047242.1 | ||||
| AK054279.1 | BAC35715.1 | |||
| AK087412.1 | ||||
| AK163663.1 | BAE37447.1 | |||
| AK173083.1 | BAD32361.1 | |||
| BY251374.1 |
More Information
Set Membership: |
|

Genomic Map