Assay Details
Target Gene Details
Entrez Gene ID: | 68369 |
Gene Name: | RIKEN cDNA 0610031O16 gene |
Gene Aliases: |
- |
Location: |
Chr.3:138210717-138239663 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610031O16Rik | NR_045760.1 | |||
| AK002733.1 |
Target Gene Details
Entrez Gene ID: | 11529 |
Gene Name: | alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide |
Gene Aliases: |
AI325182, Adh-3, Adh-3e, Adh-3t, Adh3, Adh3-e, Adh3-t, Adh4, Adt-1 |
Location: |
Chr.3:138198053-138232042 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adh7 | NM_009626.4 | 7 | 1011 | NP_033756.2 |
| AF331802.1 | 7 | 1009 | AAM22808.1 | |
| AF331803.1 | 7 | 1009 | AAM22809.1 | |
| AK009588.2 | 7 | 978 | BAE43215.1 | |
| AK009600.1 | 7 | 1024 | BAB26385.1 | |
| BC047267.1 | 7 | 940 | AAH47267.1 | |
| U20257.1 | 7 | 902 | AAA76735.1 |
More Information
Set Membership: |
|

Genomic Map