Assay Details
Target Gene Details
Entrez Gene ID: | 68032 |
Gene Name: | ER membrane protein complex subunit 4 |
Gene Aliases: |
2610318K02Rik, Tmem85 |
Location: |
Chr.2:112363019-112368027 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc4 | NM_026519.3 | NP_080795.1 | ||
| AK003668.1 | BAB22926.1 | |||
| AK012047.1 | BAB27992.1 | |||
| AK078165.1 | BAC37155.1 | |||
| AK167745.1 | BAE39782.1 | |||
| BC051926.1 | AAH51926.1 |

Genomic Map